4-Brom-3-fluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

013079-25G 013079-5G
FLUO013079-25GEA 150 EUR
FLUO013079-25G FLUO013079-5G
4-Brom-3-fluorbenzolsulfonylchlorid
4-Brom-3-fluorbenzolsulfonylchlorid
Formel: C₆H₃BrClFO₂S
Molecular Weight: 273,51 g/mol
Siedepunkt: 217…218 °C (1013 hPa)
Dichte: 1,85 g/cm³ (20 °C)
MDL: MFCD03094150
CAS-Nummer: 351003-51-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR