5-Benzyloxyindol

Lieferant: FLUOROCHEM

Synonyme: 5-Benzyloxy-1H-indol

016236-25G 016236-5G
FLUO016236-25GEA 173 EUR
FLUO016236-25G FLUO016236-5G
5-Benzyloxyindol
5-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
Siedepunkt: 165 °C (0,1 mmHg)
Schmelzpunkt: 107 °C
MDL: MFCD00005676
CAS-Nummer: 1215-59-4
EINECS: 214-930-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR