6-(Trifluormethyl)indol

Lieferant: COMBI-BLOCKS

IN-0167-5G IN-0167-1G IN-0167-25G
COBBIN-0167-5GEA 449 EUR
COBBIN-0167-5G COBBIN-0167-1G COBBIN-0167-25G
6-(Trifluormethyl)indol
6-(Trifluoromethyl)indole
Formel: C₉H₆F₃N
Molecular Weight: 185.15 g/mol
Schmelzpunkt: 109…110 °C
MDL: MFCD00272316
CAS-Nummer: 13544-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR