2-Chlor-4-(trifluormethyl)phenylboronsäure

Lieferant: FLUOROCHEM

016229-1G 016229-5G
FLUO016229-1GEA 34.1 EUR
FLUO016229-1G FLUO016229-5G
2-Chlor-4-(trifluormethyl)phenylboronsäure
2-Chlor-4-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
MDL: MFCD02684337
CAS-Nummer: 254993-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR