2-Chlor-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3833-1G BB-3833-5G BB-3833-25G
COBBBB-3833-1GEA 0 EUR
COBBBB-3833-1G COBBBB-3833-5G COBBBB-3833-25G
2-Chlor-5-(trifluormethyl)phenylboronsäure
2-Chlor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
MDL: MFCD00797335
CAS-Nummer: 182344-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR