2'-Fluor-3'-(trifluormethyl)propiophenon

Lieferant: FLUOROCHEM

006448-1G 006448-5G
FLUO006448-1GEA 34.1 EUR
FLUO006448-1G FLUO006448-5G
2'-Fluor-3'-(trifluormethyl)propiophenon
2'-Fluor-3'-(trifluormethyl)propiophenon
Formel: C₁₀H₈F₄O
Molecular Weight: 220.17 g/mol
MDL: MFCD00061192
CAS-Nummer: 207986-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR