4'-Fluor-3'-(trifluormethyl)propiophenon

Lieferant: FLUOROCHEM

006454-1G 006454-5G
FLUO006454-1GEA 31.1 EUR
FLUO006454-1G FLUO006454-5G
4'-Fluor-3'-(trifluormethyl)propiophenon
4'-Fluor-3'-(trifluormethyl)propiophenon
Formel: C₁₀H₈F₄O
Molecular Weight: 220.17 g/mol
MDL: MFCD00061264
CAS-Nummer: 239107-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR