3-Brom-2-fluor-5-methylpyridin

Lieferant: FLUOROCHEM

Synonyme: 3-Brom-2-fluor-5-picolin

012712-1G 012712-5G
FLUO012712-1GEA 31.1 EUR
FLUO012712-1G FLUO012712-5G
3-Brom-2-fluor-5-methylpyridin
3-Bromo-2-fluoro-5-methylpyridine
Formel: C₆H₅BrFN
Molecular Weight: 190,01 g/mol
MDL: MFCD03095305
CAS-Nummer: 17282-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR