2-Brom-4-fluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-4-fluorbenzolsulfonsäurechlorid, 2-Brom-4-fluorbenzolsulfochlorid

013083-5G 013083-25G
FLUO013083-5GEA 51.8 EUR
FLUO013083-5G FLUO013083-25G
2-Brom-4-fluorbenzolsulfonylchlorid
2-Brom-4-fluorbenzolsulfonylchlorid
Formel: C₆H₃BrClFO₂S
Molecular Weight: 273.51 g/mol
Siedepunkt: 264…265 °C (1013 hPa)
Dichte: 1.87 g/cm³ (20 °C)
MDL: MFCD03094149
CAS-Nummer: 351003-45-7
UN: 2920
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR