4-Brom-2,5-difluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Brom-2,5-difluorbenzolsulfonsäurechlorid, 4-Brom-2,5-difluorbenzolsulfochlorid

005237-1G 005237-5G
FLUO005237-1GEA 31.1 EUR
FLUO005237-1G FLUO005237-5G
4-Brom-2,5-difluorbenzolsulfonylchlorid
4-Brom-2,5-difluorbenzolsulfonylchlorid
Formel: C₆H₂BrClF₂O₂S
Molecular Weight: 291.5 g/mol
Siedepunkt: 76 °C (0.01 mmHg)
Schmelzpunkt: 34…36 °C
MDL: MFCD00042181
CAS-Nummer: 207974-14-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR