3-Brom-5-(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-Brom-5-(trifluormethyl)benzolsulfochlorid, 3-Brom-5-(trifluormethyl)benzol-1-sulfonylchlorid, 3-Brom-5-(trifluormethyl)benzolsulfonsäurechlorid
013085-5G
013085-1G
FLUO013085-5GEA
104
EUR
FLUO013085-5G
FLUO013085-1G
3-Brom-5-(trifluormethyl)benzolsulfonylchlorid
3-Brom-5-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃BrClF₃O₂S Siedepunkt: 217…218 °C (1013 hPa) Dichte: 1,835 g/cm³ (20 °C) |
MDL:
MFCD03094393 CAS-Nummer: 351003-46-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...