4-Brom-3-(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
4-Brom-3-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Brom-3-(trifluormethyl)benzolsulfochlorid, 4-Brom-3-(trifluormethyl)benzolsulfonsäurechlorid
013074-5G
013074-25G
013074-1G
FLUO013074-5GEA
57.6
EUR
FLUO013074-5G
FLUO013074-25G
FLUO013074-1G
4-Brom-3-(trifluormethyl)benzolsulfonylchlorid
4-Brom-3-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃BrClF₃O₂S Dichte: 1,833 g/cm³ (20 °C) |
MDL:
MFCD03094391 CAS-Nummer: 351003-47-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...