2-(Trifluormethyl)benzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluormethyl)benzolsulfonsäureamid, α,α,α-Trifluor-o-toluolsulfonamid

013873-5G 013873-25G
FLUO013873-5GEA 0 EUR
FLUO013873-5G FLUO013873-25G
2-(Trifluormethyl)benzolsulfonamid
2-(Trifluormethyl)benzolsulfonamid
Formel: C₇H₆F₃NO₂S
MDL: MFCD00042420
CAS-Nummer: 1869-24-5
UN: 2811
ADR: 6.1,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR