4-(Trifluormethyl)benzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: α,α,α-Trifluor-p-toluolsulfonamid, 4-(Trifluormethyl)benzolsulfonsäureamid

013874-5G 013874-25G
FLUO013874-5GEA 0 EUR
FLUO013874-5G FLUO013874-25G
4-(Trifluormethyl)benzolsulfonamid
4-(Trifluormethyl)benzolsulfonamid
Formel: C₇H₆F₃NO₂S
Molecular Weight: 225,19 g/mol
MDL: MFCD00159251
CAS-Nummer: 830-43-3
EINECS: 212-596-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR