3-Chlor-2-fluor-5-(trifluormethyl)benzoesäure
Lieferant: FLUOROCHEM
005679-5G
005679-1G
FLUO005679-5GEA
142
EUR
FLUO005679-5G
FLUO005679-1G
3-Chlor-2-fluor-5-(trifluormethyl)benzoesäure
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃ClF₄O₂ Molecular Weight: 242.56 g/mol |
MDL:
MFCD01631509 CAS-Nummer: 129931-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...