5-Methoxytryptamin

Lieferant: CHEMPUR

FL19999.POR 019999-2.5G
CHMPFL19999.POREA 0 EUR
CHMPFL19999.POR FLUO019999-2.5G
5-Methoxytryptamin
5-Methoxytryptamin
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,24 g/mol
MDL: MFCD00005662
CAS-Nummer: 608-07-1
EINECS: 210-153-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR