4-Benzyloxyindol

Lieferant: FLUOROCHEM

016140-1G 016140-5G
FLUO016140-1GEA 31.1 EUR
FLUO016140-1G FLUO016140-5G
4-Benzyloxyindol
4-Benzyloxyindol
Formel: C₁₅H₁₃NO
MDL: MFCD00047200
CAS-Nummer: 20289-26-3
EINECS: 243-690-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR