1-Boc-5-brom-2-indolboronsäure
Lieferant: FLUOROCHEM
Synonyme:
(5-Bromo-1-(tert-butoxycarbonyl)-1H-indol-2-yl)boronic acid, N-(Boc-5-bromo-2-indolyl)boronic acid, 5-Bromo-N-(Boc)indole-2-boronic acid
016106-1G
016106-5G
FLUO016106-1GEA
39.9
EUR
FLUO016106-1G
FLUO016106-5G
1-Boc-5-brom-2-indolboronsäure
1-Boc-5-brom-2-indolboronsäure
Formel:
C₁₃H₁₅BBrNO₄ Molecular Weight: 339.98 g/mol |
MDL:
MFCD03701683 CAS-Nummer: 475102-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...