2-Brom-4-thiazolcarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Bromo-1,3-thiazole-4-carboxylic acid, 2-Bromothiazole-4-carboxylic acid
016152-1G
016152-5G
FLUO016152-1GEA
72.9
EUR
FLUO016152-1G
FLUO016152-5G
2-Brom-4-thiazolcarbonsäure
2-Bromo-4-thiazolecarboxylic acid
Formel:
C₄H₂BrNO₂S |
MDL:
MFCD04115729 CAS-Nummer: 5198-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...