(4-Fluor-2-(trifluormethyl)phenyl)essigsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid
006439-1G
006439-5G
FLUO006439-1GEA
72.9
EUR
FLUO006439-1G
FLUO006439-5G
(4-Fluor-2-(trifluormethyl)phenyl)essigsäure
(4-Fluor-2-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆F₄O₂ Molecular Weight: 222.14 g/mol |
MDL:
MFCD00061189 CAS-Nummer: 195447-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...