1-Benzyloxy-3-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
3-Fluorobenzyloxybenzene
023291-25G
023291-5G
FLUO023291-25GEA
354
EUR
FLUO023291-25G
FLUO023291-5G
1-Benzyloxy-3-fluorbenzol
1-Benzyloxy-3-fluorbenzol
Formel:
C₁₃H₁₁FO Molecular Weight: 202,23 g/mol |
MDL:
MFCD06409983 CAS-Nummer: 72216-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...