3-Brompicolinsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid

012713-1G 012713-5G
FLUO012713-1GEA 31.1 EUR
FLUO012713-1G FLUO012713-5G
3-Brompicolinsäure
3-Brompicolinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202.01 g/mol
MDL: MFCD01320380
CAS-Nummer: 30683-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR