2-(1,1,2,2-Tetrafluorethoxy)anilin

Lieferant: Apollo Scientific

PC5839-5G PC5839-1G PC5839-25G
APOSPC5839-5GEA 95.7 EUR
APOSPC5839-5G APOSPC5839-1G APOSPC5839-25G
2-(1,1,2,2-Tetrafluorethoxy)anilin
2-(1,1,2,2-Tetrafluorethoxy)anilin
Formel: C₈H₇F₄NO
MDL: MFCD03094306
CAS-Nummer: 35295-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR