(2-Chlor-6-fluor-3-methylphenyl)essigsäure

Lieferant: FLUOROCHEM

005660-2.5G 005660-1G
FLUO005660-2.5GEA 0 EUR
FLUO005660-2.5G FLUO005660-1G
(2-Chlor-6-fluor-3-methylphenyl)essigsäure
(2-Chlor-6-fluor-3-methylphenyl)essigsäure
Formel: C₉H₈ClFO₂
Molecular Weight: 202.61 g/mol
MDL: MFCD01631365
CAS-Nummer: 261762-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR