4-Amino-3-chlor-5-nitrobenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-6-nitro-4-(trifluoromethyl)aniline

003967-1G 003967-5G
FLUO003967-1GEA 31.1 EUR
FLUO003967-1G FLUO003967-5G
4-Amino-3-chlor-5-nitrobenzotrifluorid
4-Amino-3-chlor-5-nitrobenzotrifluorid
Formel: C₇H₄ClF₃N₂O₂
MDL: MFCD00042153
CAS-Nummer: 57729-79-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR