2,2'-Dichloracetanilid

Lieferant: Thermo Scientific

KM02680DA KM02680EA
MAYBKM02680DAEA 0 EUR
MAYBKM02680DA MAYBKM02680EA
2,2'-Dichloracetanilid
2,2'-Dichloracetanilid
Formel: C₈H₇Cl₂NO
MDL: MFCD00032560
CAS-Nummer: 3289-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR