2,6-Dichlor-4-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

004744-250MG 004744-1G
FLUO004744-250MGEA 0 EUR
FLUO004744-250MG FLUO004744-1G
2,6-Dichlor-4-(trifluormethyl)benzonitril
2,6-Dichlor-4-(trifluormethyl)benzonitril
Formel: C₈H₂Cl₂F₃N
MDL: MFCD00220879
CAS-Nummer: 157021-61-9
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR