4-(1H-Pyrazol-1-yl)anilin

Lieferant: Thermo Scientific

Synonyme: 4-Pyrazol-1-yl-phenylamine

CC18414.0001 CC18414DA CC18414EA
MAYBCC18414.0001EA 0 EUR
MAYBCC18414.0001 MAYBCC18414DA MAYBCC18414EA
4-(1H-Pyrazol-1-yl)anilin
4-(1H-Pyrazol-1-yl)anilin
Formel: C₉H₉N₃
Molecular Weight: 159,1879 g/mol
MDL: MFCD00626298
CAS-Nummer: 17635-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR