2'-Amino-4'-methoxyacetanilid

Lieferant: BIONET RESEARCH

6T-0332.500 6T-0332.1 6T-0332.5 6T-0332.10
BIOT6T-0332.500EA 0 EUR
BIOT6T-0332.500 BIOT6T-0332.1 BIOT6T-0332.5 BIOT6T-0332.10
2'-Amino-4'-methoxyacetanilid
2'-Amino-4'-methoxyacetanilid
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180.2 g/mol
MDL: MFCD00026124
CAS-Nummer: 5472-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR