4-Isobutyl-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2145-5G BB-2145-1G
COBBBB-2145-5GEA 1310 EUR
COBBBB-2145-5G COBBBB-2145-1G
4-Isobutyl-3-nitrophenylboronsäure
4-Isobutyl-3-nitrophenylboronsäure
Formel: C₁₀H₁₄BNO₄
MDL: MFCD04115653
CAS-Nummer: 850568-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR