4-tert-Butyl-3-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2263-5G
BB-2263-1G
COBBBB-2263-5GEA
0
EUR
COBBBB-2263-5G
COBBBB-2263-1G
4-tert-Butyl-3-nitrophenylboronsäure
4-tert-Butyl-3-nitrophenylboronsäure
Formel:
C₁₀H₁₄BNO₄ |
MDL:
MFCD07363743 CAS-Nummer: 871329-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...