2-(Trifluormethylthio)benzylbromid

Lieferant: FLUOROCHEM

008010-1G 008010-5G
FLUO008010-1GEA 0 EUR
FLUO008010-1G FLUO008010-5G
2-(Trifluormethylthio)benzylbromid
2-(Trifluormethylthio)benzylbromid
Formel: C₈H₆BrF₃S
Molecular Weight: 271.10 g/mol
MDL: MFCD00236345
CAS-Nummer: 238403-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR