3-Chlor-4-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid
BB-2033-5G
BB-2033-25G
BB-2033-1G
COBBBB-2033-5GEA
97.9
EUR
COBBBB-2033-5G
COBBBB-2033-25G
COBBBB-2033-1G
3-Chlor-4-methylphenylboronsäure
3-Chlor-4-methylphenylboronsäure
Formel:
C₇H₈BClO₂ Molecular Weight: 170,40 g/mol |
MDL:
MFCD04039010 CAS-Nummer: 175883-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...