4-Chlor-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-2-methylbenzeneboronic acid

BB-2189-5G BB-2189-25G
COBBBB-2189-5GEA 43.5 EUR
COBBBB-2189-5G COBBBB-2189-25G
4-Chlor-2-methylphenylboronsäure
4-Chlor-2-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170.40 g/mol
MDL: MFCD02683107
CAS-Nummer: 209919-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR