4-Chlor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-3-methylbenzeneboronic acid

BB-2034-1G BB-2034-5G
COBBBB-2034-1GEA 50.7 EUR
COBBBB-2034-1G COBBBB-2034-5G
4-Chlor-3-methylphenylboronsäure
4-Chlor-3-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170,40 g/mol
MDL: MFCD02683104
CAS-Nummer: 161950-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR