2-Methoxy-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5277-5G BB-5277-1G
COBBBB-5277-5GEA 1330 EUR
COBBBB-5277-5G COBBBB-5277-1G
2-Methoxy-3-(trifluormethyl)phenylboronsäure
2-Methoxy-3-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₃
MDL: MFCD07772004
CAS-Nummer: 1072946-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR