2-Methoxy-5-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methoxy-5-(trifluoromethyl)benzeneboronic acid
BB-2789-1G
BB-2789-5G
COBBBB-2789-1GEA
58.4
EUR
COBBBB-2789-1G
COBBBB-2789-5G
2-Methoxy-5-(trifluormethyl)phenylboronsäure
2-Methoxy-5-(trifluormethyl)phenylboronsäure
Formel:
C₈H₈BF₃O₃ Molecular Weight: 219.95 g/mol |
MDL:
MFCD04039222 CAS-Nummer: 240139-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...