2-Propoxy-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6364-5G BB-6364-1G
COBBBB-6364-5GEA 0 EUR
COBBBB-6364-5G COBBBB-6364-1G
2-Propoxy-5-(trifluormethyl)phenylboronsäure
2-Propoxy-5-(trifluormethyl)phenylboronsäure
Formel: C₁₀H₁₂BF₃O₃
MDL: MFCD16036114
CAS-Nummer: 1162257-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR