3-Ethoxy-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Ethoxy-5-trifluoromethylphenylboronic acid

BB-2848-5G BB-2848-1G
COBBBB-2848-5GEA 742 EUR
COBBBB-2848-5G COBBBB-2848-1G
3-Ethoxy-5-(trifluormethyl)phenylboronsäure
3-Ethoxy-5-(trifluormethyl)phenylboronsäure
Formel: C₉H₁₀BF₃O₃
Molecular Weight: 233.98 g/mol
MDL: MFCD08056361
CAS-Nummer: 871332-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR