3-Methoxy-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Methoxy-5-trifluoromethylphenylboronic acid

BB-2849-5G BB-2849-1G
COBBBB-2849-5GEA 380 EUR
COBBBB-2849-5G COBBBB-2849-1G
3-Methoxy-5-(trifluormethyl)phenylboronsäure
3-Methoxy-5-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219.95 g/mol
MDL: MFCD08056362
CAS-Nummer: 871332-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR