4-Isopropoxy-2-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Isopropoxy-2-trifluoromethylphenylboronic acid

BB-2266-1G BB-2266-250M
COBBBB-2266-1GEA 312 EUR
COBBBB-2266-1G COBBBB-2266-250M
4-Isopropoxy-2-(trifluormethyl)phenylboronsäure
4-Isopropoxy-2-(trifluormethyl)phenylboronsäure
Formel: C₁₀H₁₂BF₃O₃
MDL: MFCD04972373
CAS-Nummer: 313545-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR