4-Ethoxy-2-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Ethoxy-2-(trifluoromethyl)phenyl)boronic acid
BB-3289-1G
BB-3289-250M
COBBBB-3289-1GEA
474
EUR
COBBBB-3289-1G
COBBBB-3289-250M
4-Ethoxy-2-(trifluormethyl)phenylboronsäure
4-Ethoxy-2-(trifluormethyl)phenylboronsäure
Formel:
C₉H₁₀BF₃O₃ Molecular Weight: 233,98 g/mol |
MDL:
MFCD08059515 CAS-Nummer: 313545-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...