4-Ethoxy-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Ethoxy-3-(trifluoromethyl)benzeneboronic acid, 4-Ethoxy-3-trifluoromethylphenylboronic acid

BB-2795-5G BB-2795-1G
COBBBB-2795-5GEA 1140 EUR
COBBBB-2795-5G COBBBB-2795-1G
4-Ethoxy-3-(trifluormethyl)phenylboronsäure
4-Ethoxy-3-(trifluormethyl)phenylboronsäure
Formel: C₉H₁₀BF₃O₃
Molecular Weight: 233,98 g/mol
MDL: MFCD07363789
CAS-Nummer: 871329-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR