4-Methoxy-2-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxy-2-(trifluoromethyl)benzeneboronic acid, (4-Methoxy-2-(trifluoromethyl)phenyl)boronic acid

BB-8402-5G BB-8402-1G
COBBBB-8402-5GEA 497 EUR
COBBBB-8402-5G COBBBB-8402-1G
4-Methoxy-2-(trifluormethyl)phenylboronsäure
4-Methoxy-2-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219,95 g/mol
MDL: MFCD03095346
CAS-Nummer: 313546-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR