4-Methoxy-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methoxy-3-(trifluoromethyl)phenyl)boronic acid, 4-Methoxy-3-trifluoromethylphenylboronic acid, 4-Methoxy-3-(trifluoromethyl)benzeneboronic acid

BB-2796-5G BB-2796-1G
COBBBB-2796-5GEA 531 EUR
COBBBB-2796-5G COBBBB-2796-1G
4-Methoxy-3-(trifluormethyl)phenylboronsäure
4-Methoxy-3-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219.95 g/mol
MDL: MFCD07363790
CAS-Nummer: 149507-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR