N-Ethyl-2-nitro-4-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

AN-1255-5G AN-1255-1G
COBBAN-1255-5GEA 303 EUR
COBBAN-1255-5G COBBAN-1255-1G
N-Ethyl-2-nitro-4-(trifluormethyl)anilin
N-Ethyl-2-nitro-4-(trifluormethyl)anilin
Formel: C₉H₉F₃N₂O₂
MDL: MFCD00044178
CAS-Nummer: 30377-62-9
EINECS: 250-160-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR