N-Methyl-2-nitro-4-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Methylamino)-3-nitrobenzotrifluoride

AN-1257-5G AN-1257-1G AN-1257-25G
COBBAN-1257-5GEA 149 EUR
COBBAN-1257-5G COBBAN-1257-1G COBBAN-1257-25G
N-Methyl-2-nitro-4-(trifluormethyl)anilin
N-Methyl-2-nitro-4-(trifluormethyl)anilin
Formel: C₈H₇F₃N₂O₂
Molecular Weight: 220,1486 g/mol
MDL: MFCD00798375
CAS-Nummer: 20200-22-0
EINECS: 243-586-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR