Tetrathiafulvalen ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2'-Bi(1,3-dithiolylidene), TTF
L19229.03
L19229.MD
L19229.03EA
150
EUR
L19229.03
L19229.MD
Tetrathiafulvalen ≥97%
Tetrathiafulvalen
Purity: 97%
CAS No.: 31366-25-3
Molecular Formula: C6H4S4
Formula Weight: 204.35
Melting Point: 116-120°
Beilstein Reference: 1617956
Merck Reference: 14,9242
MDL No.: MFCD00005492
Formel:
C₆H₄S₄ Molecular Weight: 204,356 g/mol Schmelzpunkt: 116…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005492 CAS-Nummer: 31366-25-3 EINECS: 250-593-7 Merck Index: 14,9242 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...