Tetrathiafulvalen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-Bi(1,3-dithiolylidene), TTF

M-Clarity™ Qualitätsstufe = MQ200

183180-1G 183180-250MG
SIAL183180-1GEA 423 EUR
SIAL183180-1G SIAL183180-250MG
Tetrathiafulvalen, Sigma-Aldrich®
Tetrathiafulvalen
Formel: C₆H₄S₄
Molecular Weight: 204.356 g/mol
MDL: MFCD00005492
CAS-Nummer: 31366-25-3
EINECS: 250-593-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR