Furfuryldisulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Bis(furan-2-ylmethyl)disulphane

M-Clarity™ Qualitätsstufe = MQ300

W314609-100G-K W314609-1KG-K W314609-250G-K
SIALW314609-100G-KEA 185 EUR
SIALW314609-100G-K SIALW314609-1KG-K SIALW314609-250G-K
Furfuryldisulfid, Sigma-Aldrich®
Furfuryldisulfid
Formel: C₁₀H₁₀O₂S₂
Molecular Weight: 226,3152 g/mol
MDL: MFCD00010082
CAS-Nummer: 4437-20-1
EINECS: 224-649-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR